Sodium trichloroacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

184150025 184151000 184155000
TS184150025EA 369 USD
TS184150025 TS184151000 TS184155000
Sodium trichloroacetate 97%
Sodium trichloroacetate
Formula: C₂Cl₃NaO₂
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064198
CAS Number: 650-51-1
EINECS: 211-479-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Powder or granular powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %
Loss on drying ≤1 %
Solubility (1 % in methanol) Clear colorless to yellow solution
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR