Ethanol-[D₁] 99.5+ atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

166320250 166321000
TS166320250EA 84 USD
TS166320250 TS166321000
Ethanol-[D₁] 99.5+ atom % D for NMR spectroscopy
Ethanol-[D₁]
Formula: C₂H₅DO
Boiling Pt: 78 °C (1013 hPa)
Melting Pt: –114.5 °C
Density: 0.8 g/cm³
Flash Pt: 12 °C
CAS Number: 925-93-9
EINECS: 213-128-9
UN: 1170

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.5 % (Coulometric)
D-Enrichment ≥99.50 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR