Acetic acid-[D₄] (99.5% D) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethanoic acid-D4, Acetic-D3 acid-D, Acetic-d3 acid-d, Acetic acid-[²H₄], Tetradeuteroacetic acid, Acetic acid-d₄
166210250
166210500
166210050
TS166210250EA
190
USD
TS166210250
TS166210500
TS166210050
Acetic acid-[D₄] (99.5% D) for NMR spectroscopy
Acetic acid-[D4]
Formula:
D₃CCOOD MW: 64.02 g/mol Boiling Pt: 115.5 °C (760 mmHg) Melting Pt: 15…16 °C Density: 1.13 g/cm³ Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051051 CAS Number: 1186-52-3 EINECS: 214-693-4 UN: 2789 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.1 % (Coulometric) |
D-Enrichment | ≥99.4 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...