Sodium trifluoroacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 2,2,2-trifluoroacetate

157930250 157931000
TS157930250EA 40 USD
TS157930250 TS157931000
Sodium trifluoroacetate 97%
Sodium trifluoroacetate
Formula: C₂F₃NaO₂
MW: 136.01 g/mol
Melting Pt: 207…209 °C
MDL Number: MFCD00013217
CAS Number: 2923-18-4
EINECS: 220-879-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 205 to 207 °C
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR