Tetraethylammonium hydroxide 20% (w/w) in water
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetrylammonium hydroxide
176072500
176070010
176070050
TS176072500EA
97
USD
TS176072500
TS176070010
TS176070050
Tetraethylammonium hydroxide 20% (w/w) in water
Tetraethylammonium hydroxide
Formula:
C₈H₂₁NO MW: 147.26 g/mol Density: 1.02 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00009024 CAS Number: 77-98-5 EINECS: 201-073-3 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Solution |
Titration with HCl | ≥20 % |
GC | ≥96.0 % |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...