Dibutyl ether 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-n-butyl ether

149690010 149690025 149690250
TS149690010EA 106 USD
TS149690010 TS149690025 TS149690250
Dibutyl ether 99+%, extra pure
Dibutyl ether
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 141
Melting Pt: -95
Density: 0.768 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00009461
CAS Number: 142-96-1
EINECS: 205-575-3
UN: 1149
ADR: 3 III
Merck Index: 13,01568

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3980 to 1.4000 (20°C, 589 nm)
Water ≤0.05 %
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.01 %
Peroxides ≤0.01 % dibutylperoxide


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR