Methanesulfonic acid 70% in water, pure

Supplier: Thermo Scientific Chemicals
Danger

125620050 125622500 125620010
TS125620050EA 211 USD
TS125620050 TS125622500 TS125620010
Methanesulfonic acid 70% in water, pure
Methanesulphonic acid
Formula: CH₃SO₃H
MW: 96.11 g/mol
Melting Pt: –54 °C
Density: 1.35 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤25 APHA
Titration with NaOH 69.5 to 70.5 %
Chloride (Cl) ≤25 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR