Bis(2-ethoxyethyl)ether 98+%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Ethoxyethyl ether, Diethyleneglycoldiethylether, Diethylene glycol diethyl ether

117900010 117900025 117900100 117905000
TS117900010EA 155 USD
TS117900010 TS117900025 TS117900100 TS117905000
Bis(2-ethoxyethyl)ether 98+%, extra pure
Bis(2-ethoxyethyl)ether
Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Boiling Pt: 189
Melting Pt: -44
Density: 0.9 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00009254
CAS Number: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4130 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥98.0 %
Color scale ≤15 APHA
Residue after evaporation ≤0.05 %
Acidity (CH3COOH) ≤0.015 %
Peroxides ≤0.0015 % (as H2O2)
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR