N,N-Dimethylacetamide 99+%, extra_pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
181070250
181070025
181070050
TS181070250EA
1137
USD
TS181070250
TS181070010
TS181070025
TS181070050
N,N-Dimethylacetamide 99+%, extra_pure
N,N-Dimethylacetamide
Formula:
CH₃CON(CH₃)₂ MW: 87.12 g/mol Boiling Pt: 166.1 Melting Pt: -20 Density: 0.937 g/cm³ Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008686 CAS Number: 127-19-5 EINECS: 204-826-4 Merck Index: 13,03254 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4370 to 1.4390 (20°C, 589 nm) |
Water | ≤0.1 % |
GC | ≥99.0 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.01 % |
Free acid (CH3COOH) | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...