Nitronium tetrafluoroborate 0.5 M in sulfolane

Supplier: Thermo Scientific Chemicals
Danger

175130250 175132000
TS175130250EA 110 USD
TS175130250 TS175132000
Nitronium tetrafluoroborate 0.5 M in sulfolane
Nitronium tetrafluoroborate
Formula: BF₄NO₂
Density: 1.268 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011432
CAS Number: 13826-86-3
EINECS: 237-533-5
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to yellow Solution
Strength 0.48 to 0.55 M
Melting point 88 to 92 °C
Loss on drying ≤0.5 % (1 g, 60 °C, 4 h)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR