Nitric acid 60% water, extra_pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nitric acid solution

133620011 133620025 133620026 133620010
TS133620011EA 91 USD
TS133620011 TS133620025 TS133620026 TS133620010
Nitric acid 60% water, extra_pure
Nitric acid
Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.33 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Titration with NaOH 59.0 to 63.0 %
Chloride (Cl) ≤10 ppm
Iron (Fe) ≤5 ppm
NMR (31P-NMR) Consistent with structure
Elemental Analysis 7.6 to 8.2 % H
Specific optical rotation +78° to +86° (20°C, 589 nm) (c=0.8 in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR