Chlorobenzene 99+%, pure

Supplier: Thermo Scientific Chemicals
Warning

146410010 146410025 146410250
TS146410010EA 108 USD
TS146410010 TS146410025 TS146410250
Chlorobenzene 99+%, pure
Chlorobenzene
Formula: C₆H₅Cl
Boiling Pt: 131 °C (760 mmHg)
Melting Pt: –45 °C
Density: 1.1075 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5230 to 1.5250 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR