1,2,4-Trichlorobenzene 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

157900010 157900025 157900100
TS157900010EA 124 USD
TS157900010 TS157900025 TS157900100
1,2,4-Trichlorobenzene 99%, pure
1,2,4-Trichlorobenzene
Formula: C₆H₃Cl₃
Boiling Pt: 214 °C (760 mmHg)
Melting Pt: 16 °C
Density: 1.45 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5700 to 1.5720 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤30 APHA
Sulfate (SO4) ≤0.002 %
Ammonium (NH4) ≤0.002 %
Iron (Fe) ≤0.001 %
Potassium (K) ≤0.005 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.005 %
Neutrality Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR