5-Methyl-2-hexanone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Isoamyl methyl ketone, Methyl isoamyl ketone
149660010
149660025
TS149660010EA
110
USD
TS149660010
TS149660025
5-Methyl-2-hexanone 99%
5-Methyl-2-hexanone
Formula:
C₇H₁₄O Boiling Pt: 144 °C (1013 hPa) Melting Pt: –74 °C Density: 0.81 g/cm³ Flash Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008950 CAS Number: 110-12-3 EINECS: 203-737-8 UN: 2302 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4060 to 1.4090 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific optical rotation | +16° to +23° (20°C, 589 nm) (c=1 in H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...