Methanol 99.9% analysis_for
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
176840010
176840025
176840100
176840250
176845000
TS176840010EA
46
USD
TS176840010
TS176840025
TS176840100
TS176840250
TS176845000
Methanol 99.9% analysis_for
Methanol
Formula:
CH₃OH MW: 32.04 g/mol Boiling Pt: 64.6 Melting Pt: -98 Density: 0.791 g/cm³ Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3 II REACH: 01-2119433307-44 Merck Index: 13,05984 |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.05 % (K.F.) |
Solubility | (in water) Passes test |
GC | ≥99.85 % |
Color scale | ≤5 APHA |
Residue after evaporation | ≤0.001 % |
Acidity/alkalinity | ≤0.0005 meq/g |
Substance darkened by H2SO4 | ≤5 APHA |
Calcium (Ca) | ≤0.2 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.02 ppm |
Magnesium (Mg) | ≤0.05 ppm |
Potassium (K) | ≤0.5 ppm |
Sodium (Na) | ≤0.5 ppm |
Zinc (Zn) | ≤0.05 ppm |
Carbonyl compounds | ≤0.001 % |
E/E | ≤0.05 % |
2-Propanol | ≤0.005 % |
KMnO4-reducing substance | ≤0.00025 % |
Total phosphorus (P) | ≤0.05 ppm |
Total silicon (Si) | ≤0.05 ppm |
Total sulfur (S) | ≤0.1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...