1-Propanol 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

149480010 149480025 149480250
TS149480010EA 70 USD
TS149480010 TS149480025 TS149480250
1-Propanol 99+%, Extra Pure
1-Propanol
Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97 °C (760 mmHg)
Melting Pt: –127 °C
Density: 0.8 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3840 to 1.3860 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR