1-Butanol 99+% for spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
167690025
167691000
167695000
TS167690025EA
154
USD
TS167690025
TS167691000
TS167695000
1-Butanol 99+% for spectroscopy
1-Butanol
Formula:
H₃C(CH₂)₂CH₂OH Boiling Pt: 117.6 °C (760 mmHg) Melting Pt: –89 °C Density: 0.81 g/cm³ Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III REACH: 01-2119484630-38 |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.05 % (Coulometric) |
GC | ≥99.0 % |
UV | 1 cm cell vs HPLC-grade water |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...