1,3-Dichloropropane 99%

Supplier: Thermo Scientific Chemicals
Danger

113680500 113682500
TS113680500EA 143 USD
TS113680500 TS113682500
1,3-Dichloropropane 99%
1,3-Dichloropropane
Formula: ClH₂CCH₂CH₂Cl
Boiling Pt: 120…122 °C (760 mmHg)
Melting Pt: –99 °C
Density: 1.19 g/cm³
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00000999
CAS Number: 142-28-9
EINECS: 205-531-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4471 to 1.4491 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR