Isoamyl acetate 99+%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopentyl acetate, Acetic acid isoamyl ester

150660025 150662500 150660010
TS150660025EA 143 USD
TS150660025 TS150662500 TS150660010
Isoamyl acetate 99+%, pure
Isoamyl acetate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 142 °C (756 mmHg)
Melting Pt: –78 °C
Density: 0.874 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008946
CAS Number: 123-92-2
EINECS: 204-662-3
UN: 1104
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4030 (20°C, 589 nm)
GC ≥98.5 % (sum of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR