3-Pentanone 98%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethyl ketone

155810025 155815000
TS155810025EA 173 USD
TS155810025 TS155815000
3-Pentanone 98%, pure
3-Pentanone
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 100…102 °C (760 mmHg)
Melting Pt: –40 °C
Density: 0.815 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009320
CAS Number: 96-22-0
EINECS: 202-490-3
UN: 1156
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3930 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR