Acetonitrile 99+% Spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
167650010
167650025
167652500
TS167650010EA
184
USD
TS167650010
TS167650025
TS167652500
Acetonitrile 99+% Spectroscopy
Acetonitrile
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 81.6 Melting Pt: -45.7 Density: 0.781 g/cm³ Flash Pt: 2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3 II REACH: 01-2119471307-38 Merck Index: 13,00071 |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.03 % |
GC | ≥99.0 % |
UV | at 250 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...