1,2,3,4-Tetrahydronaphthalene 98+%, pure

Supplier: Thermo Scientific Chemicals
Warning

146730010 146730025 146730100
TS146730010EA 120 USD
TS146730010 TS146730025 TS146730100
1,2,3,4-Tetrahydronaphthalene 98+%, pure
1,2,3,4-Tetrahydronaphthalene
Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 207 °C (760 mmHg)
Melting Pt: –35 °C
Density: 0.973 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001733
CAS Number: 119-64-2
EINECS: 204-340-2
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR