Cyclohexane-[D₁₂] 99.5 atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
166280051
166280251
TS166280051EA
135
USD
TS166280051
TS166280251
Cyclohexane-[D₁₂] 99.5 atom % D for NMR spectroscopy
Cyclohexane-[D₁₂]
Formula:
C₆D₁₂ |
MDL Number:
MFCD00044212 CAS Number: 1735-17-7 |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.03 % |
D-Enrichment | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...