Chlorobenzene-[D₅] 98.5+ atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

175320010 175320050
TS175320010EA 56 USD
TS175320010 TS175320050
Chlorobenzene-[D₅] 98.5+ atom % D for NMR spectroscopy
Chlorobenzene-[D₅]
Formula: C₆D₅Cl
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –46 °C
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00000531
CAS Number: 3114-55-4
EINECS: 221-482-0
UN: 1134
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
D-Enrichment ≥98.50 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR