Acetone-[D₆] 99.5 atom % D + 1% TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
181160100
181160500
TS181160100EA
57
USD
TS181160100
TS181160500
Acetone-[D₆] 99.5 atom % D + 1% TMS for NMR spectroscopy
Acetone-[D₆]
Formula:
D₃CC(O)CD₃ Boiling Pt: 55.5 °C (1013 hPa) Melting Pt: –93 °C Density: 0.87 g/cm³ Flash Pt: –20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00044635 CAS Number: 666-52-4 EINECS: 211-563-9 UN: 1090 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.05 % (Coulometric) |
D-Enrichment | ≥99.4 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...