Amberlite® HPR900 (OH) ion-exchange resin

Supplier: Thermo Scientific Chemicals
Danger

301340025 301341000 301345000
TS301340025EA 255 USD
TS301340025 TS301341000 TS301345000
Amberlite® HPR900 (OH) ion-exchange resin
Amberlite® HPR900 (OH)
Storage Temperature: Ambient
MDL Number: MFCD00802695
CAS Number: 197809-12-4

Order Now

Specification Test Results

Appearance Cream Beads
Particle size ≤5.0 % (> 1.180 mm)
Harmonic mean size 0.560 to 0.700 mm
Uniformity coefficient ≤1.45
Moisture holding capacity 66.0 to 75.0 % (OH)
Total volume capacity ≥0.9 eq/l (OH)
Impurity ≤1.0 % Chloride sites content


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR