Amberlite® IRC-120, H form, ion-exchange resin
Supplier: Thermo Scientific Chemicals
|
Danger
|
466710050
466712500
TS466710050EA
247
USD
TS466710050
TS466712500
Amberlite® IRC-120, H form, ion-exchange resin
Amberlite® IRC-120, H form, ion-exchange resin
- Inorganic
- for organic synthesis
Storage Temperature:
Ambient |
MDL Number:
MFCD00132707 CAS Number: 65098-55-7 |
Specification Test Results
Appearance | Amber to light brown Beads |
Particle size | ≤4.0 % (> 1180um) |
Moisture holding capacity | 48.0 to 58.0 (H+) |
Total volume capacity | ≥1.80 eq/L (H+) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...