Amberlite® XAD4

Supplier: Thermo Scientific Chemicals

202230025 202231000 202235000
TS202230025EA 528 USD
TS202230025 TS202231000 TS202235000
Amberlite® XAD4
Amberlite® XAD4
Storage Temperature: Ambient
MDL Number: MFCD00132704
CAS Number: 9003-69-4

Order Now

Specification Test Results

Appearance White to yellow Beads
Particle size ≤5.0 % (> 1.180 mm)
Harmonic mean size 0.490 to 0.690 mm
Uniformity coefficient ≤2.0
Moisture holding capacity 54.0 to 60.0 %
Ponceau dye test ≤1.0
Sodium chloride (NaCl) 20.0 to 32.0 g/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR