Amberlite® CG-50 ion-exchange resin Type 1

Supplier: Thermo Scientific Chemicals

202110250 202111000 202115000
TS202110250EA 73 USD
TS202110250 TS202111000 TS202115000
Amberlite® CG-50 ion-exchange resin Type 1
Amberlite® CG-50
Storage Temperature: Ambient
MDL Number: MFCD00132693
CAS Number: 9042-11-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size ≤10 % (> 150 µm)
Moisture ≤10 %
Total weight capacity ≥10 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR