Amberlite® IRA-900 (Cl) ion-exchange resin
Supplier: Thermo Scientific Chemicals
202310025
202311000
202315000
TS202310025EA
249
USD
TS202310025
TS202311000
TS202315000
Amberlite® IRA-900 (Cl) ion-exchange resin
Amberlite® IRA-900 (Cl)
Storage Temperature:
Ambient |
MDL Number:
MFCD00132712 CAS Number: 9050-97-9 |
Specification Test Results
Appearance | Cream to beige to light brown Beads |
Particle size | ≤3.0 % (> 1.180 mm) |
Harmonic mean size | 0.650 to 0.820 mm |
Uniformity coefficient | ≤1.8 |
Moisture holding capacity | 58 to 64 % (Cl-) |
Total volume capacity | ≥1.0 eq/l (Cl-) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...