Petroleum spirit, 40…60 °C, ECD tested for residue analysis lindane/DDT <5ng/l, res <0.0003%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

326730010 326730025
TS326730010EA 58 USD
TS326730010 TS326730025
Petroleum spirit, 40…60 °C, ECD tested for residue analysis lindane/DDT <5ng/l, res <0.0003%
Petroleum spirit

ECD tested lindane/DDT<5ng/l, res <0.0003%

Boiling Pt: 40…60 °C (1013 hPa)
Density: 0.65 g/cm³
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.005 % (Coulometric)
Boiling point 40 to 60 °C
Color scale ≤10 APHA
GC/ECD (as lindane) ≤5 ng/l
Residue after evaporation ≤0.0003 %
Acidity (CH3COOH) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR