Petroleum spirit, 40…60 °C, ECD tested halocarbons free grade for residue analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gasoline, Petroleum ether, Petrol, Benzine
326720010
326720025
TS326720010EA
60
USD
TS326720010
TS326720025
Petroleum spirit, 40…60 °C, ECD tested halocarbons free grade for residue analysis
Petroleum spirit
ECD tested halocarbons free.
Boiling Pt:
40…60 °C (1013 hPa) Density: 0.65 g/cm³ Flash Pt: –45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 265-151-9 UN: 1268 ADR: 3,II |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.005 % (Coulometric) |
Boiling point | 40 to 60 °C |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0002 % |
Acidity (CH3COOH) | ≤0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...