Zinc chloride 1.0 M in diethyl ether, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

370051000 370058000
TS370051000EA 66 USD
TS370051000 TS370058000
Zinc chloride 1.0 M in diethyl ether, AcroSeal®
Zinc chloride
Formula: ZnCl₂
Flash Pt: –21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2924
ADR: 3,I

Order Now

Specification Test Results

Appearance Colorless to light yellow to light blue-grey Solution (May form two phases which can recombine on cooling. May develop some turbidity or precipitate)
Titration Argentometric 0.9 to 1.1 M
Water ≤0.2 % (Coulometric)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR