Periodic acid 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

198400025 198401000 198405000
TS198400025EA 1459 USD
TS198400025 TS198401000 TS198405000
Periodic acid 99+%, extra pure
Periodic acid
Formula: H₅IO₆
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Titration Iodometric ≥99.0 %
Sulfated ash ≤0.05 %
Identification Positive for periodate
Iodide (I) ≤10 ppm
Bromide and Bromate (as Cl) ≤200 ppm
Chloride and Chlorate (as Cl) ≤200 ppm
Nitrate (NO3) ≤100 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤50 ppm
Insoluble matter ≤0.01 % (in water)
Iodic acid ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR