n-Heptane 99+% for residue analysis, for trace analysis of polyaromatic hydrocarbons

Supplier: Thermo Scientific Chemicals
Danger

326770010 326770025
TS326770010EA 132 USD
TS326770010 TS326770025
n-Heptane 99+% for residue analysis, for trace analysis of polyaromatic hydrocarbons
n-Heptane
Formula: H₃C(CH₂)₅CH₃
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –91 °C
Density: 0.683 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.001 % (Coulometric)
GC ≥99 %
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %
Acidity ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR