Hexane (mixture of isomers) 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexanes, Isohexane

411540010 411540025 411540100
TS411540010EA 82 USD
TS411540010 TS411540025 TS411540100
Hexane (mixture of isomers) 99%, pure
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3810 (20°C, 589 nm)
GC ≥98.5 % (sum of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR