10H-Phenothiazine-10-propanesulfonic acid sodium salt

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 3-(10H-phenothiazin-10-yl)propane-1-sulphonate

464000010 464000050
TS464000010EA 131 USD
TS464000010 TS464000050
10H-Phenothiazine-10-propanesulfonic acid sodium salt
10H-Phenothiazine-10-propanesulfonic acid sodium salt

  • Bioactive

Formula: C₁₅H₁₄NNaO₃S₂
Storage Temperature: Refrigerator
MDL Number: MFCD08275674
CAS Number: 101199-38-6

Order Now

Specification Test Results

Appearance Almost white to grey to brown Powder
Infrared spectrum Conforms
Titration 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR