(Trimethylsilyl)methyllithium 10% (w/w) 0.7 M in hexanes, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lithium,[(trimethylsilyl)methyl]-, lithium, (trimethylsilyl)methyl-, lithiomethyltrimethylsilane

377458000 377451000
TS377458000EA 854 USD
TS377458000 TS377451000
(Trimethylsilyl)methyllithium 10% (w/w) 0.7 M in hexanes, AcroSeal®
(Trimethylsilyl)methyllithium
Formula: C₄H₁₁LiSi
MW: 94.157 g/mol
Boiling Pt: 65…70 °C
Density: 0.65 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00010747
CAS Number: 1822-00-0
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to yellow-brown Solution
Assay 7 to 14 % w/w

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR