Chloro(1,5-hexadiene)rhodium(I) dimer

Supplier: Thermo Scientific Chemicals
Warning

377920010 377922500
TS377920010EA 588 USD
TS377920010 TS377922500
Chloro(1,5-hexadiene)rhodium(I) dimer
Chloro(1,5-hexadiene)rhodium(I) dimer

Appearance: Golden Crystalline powder

Formula: C₁₂H₂₀Cl₂Rh₂
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00064595
CAS Number: 32965-49-4

Order Now

Specification Test Results

Appearance Golden Crystalline powder
Infrared spectrum Conforms
Rhodium (Rh) ≥45.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR