Sulfur trioxide N,N-dimethylformamide complex 47+% (active SO₃)

Supplier: Thermo Scientific Chemicals
Danger

425850050 425850250
TS425850050EA 60 USD
TS425850050 TS425850250
Sulfur trioxide N,N-dimethylformamide complex 47+% (active SO₃)
Sulfur trioxide N,N-dimethylformamide complex
Formula: C₃H₇NO₄S
Melting Pt: 155…158 °C
Flash Pt: >120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043411
CAS Number: 29584-42-7
EINECS: 249-701-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Beige to light brown Crystals
Infrared spectrum Conforms
Titration with NaOH ≥47.0 % (as SO3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR