1,1-Di(tert-butylperoxy)cyclohexane 50% in mineral oil

Supplier: Thermo Scientific Chemicals
Danger

361310100 361312500
TS361310100EA 31 USD
TS361310100 TS361312500
1,1-Di(tert-butylperoxy)cyclohexane 50% in mineral oil
1,1-Di(tert-butylperoxy)cyclohexane

Appearance: Clear liquid

Formula: C₁₄H₂₈O₄
MW: 260.374 g/mol
Boiling Pt: 52…54 °C (.1 mmHg)
Density: 0.95 g/cm³
Storage Temperature: Refrigerator
CAS Number: 3006-86-8
UN: 3105
ADR: 5.2,

Order Now

Specification Test Results

Appearance Clear liquid
Titration Iodometric 49 to 51 %
Color scale ≤20 APHA
Impurity ≤0.2 % Hydroperoxides (as TBHP)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR