Sodium-tert-pentyloxide 30% in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tert-pentoxide, Sodium tert-pentoxide, 95%, tert-Amyl alcohol sodium salt

252531000 252538000
TS252531000EA 73 USD
TS252531000 TS252538000
Sodium-tert-pentyloxide 30% in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Sodium-tert-pentyloxide
Formula: C₅H₁₁NaO
MW: 110.132 g/mol
Density: 0.91 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00064245
CAS Number: 14593-46-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Yellow to orange to brown Solution (May develop some turbidity or precipitate)
Titration with HClO4 ≥30 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR