Trimethylsulfonium hydroxide 0.25 M in methanol

Supplier: Thermo Scientific Chemicals
Danger

381360010 381360100
TS381360010EA 99 USD
TS381360010 TS381360100
Trimethylsulfonium hydroxide 0.25 M in methanol
Trimethylsulfonium Hydroxide
Formula: C₃H₁₀OS
Density: 0.81 g/cm³
Flash Pt: 11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216756
CAS Number: 17287-03-5
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl 0.20 to 0.30 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR