Hexadecyltrimethylammonium hydroxide 25% in methanol

Supplier: Thermo Scientific Chemicals
Danger

411420500 411421000
TS411420500EA 242 USD
TS411420500 TS411421000
Hexadecyltrimethylammonium hydroxide 25% in methanol
Hexadecyltrimethylammonium hydroxide

Appearance: Clear colorless to light yellow Solution

Formula: C₁₉H₄₃NO
Density: 0.83 g/cm³
Flash Pt: 11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041921
CAS Number: 505-86-2
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Water ≤3 % (K.F.)
Concentration 23 to 27 w/w% (in methanol)
Enantiomeric excess ≥97.5 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR