Eplerenone
Supplier: Thermo Scientific Chemicals
Synonyms:
9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic Acid gamma-Lactone Methyl Ester
462970010
462972500
TS462970010EA
771
USD
TS462970010
TS462972500
Eplerenone
Eplerenone
- Bioactive
Formula:
C₂₄H₃₀O₆ MW: 414.50 g/mol Melting Pt: 242 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD07783647 CAS Number: 107724-20-9 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
Sulfated ash | ≤0.1 % |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...