Tetrakis(hydroxymethyl)phosphonium chloride ∼75-85% in water

Supplier: Thermo Scientific Chemicals
Danger

389441000 389448000
TS389441000EA 63 USD
TS389441000 TS389448000
Tetrakis(hydroxymethyl)phosphonium chloride ∼75-85% in water
Tetrakis(hydroxymethyl)phosphonium chloride
Formula: C₄H₁₂ClO₄P
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00031687
CAS Number: 124-64-1
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to yellow-green Solution
Infrared spectrum Conforms
Titration Argentometric 75 to 85 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR