Boron trifluoride acetic acid complex (1:2) (∼33% BF₃)

Supplier: Thermo Scientific Chemicals
Danger

277220250 277225000
TS277220250EA 35 USD
TS277220250 TS277225000
Boron trifluoride acetic acid complex (1:2) (∼33% BF₃)
Boron trifluoride acetic acid complex (1:2)
Formula: C₄H₈BF₃O₄
Boiling Pt: 140…148 °C (760 mmHg)
Melting Pt: –47 °C
Density: 1.345 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00036359
CAS Number: 373-61-5
EINECS: 206-768-5
UN: 1742
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow to yellow brown Liquid (May darken on storage)
Assay 33 to 37 % (BF3)
Free acid (CH3COOH) 63 to 67 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR