Hexafluorosilicic acid 25% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

410710025 410715000
TS410710025EA 144 USD
TS410710025 TS410715000
Hexafluorosilicic acid 25% (w/w) in aqueous solution
Hexafluorosilicic acid
Formula: H₂F₆Si
Density: 1.27 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00036289
CAS Number: 16961-83-4
EINECS: 241-034-8
UN: 1778
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Assay 19 to 20.6 % (total fluorine)
GC ≥94 %
Specific optical rotation +24° to +28° (20°C, 589 nm) (C=1, Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR