N-Methylmorpholine-N-oxide 50% (w/w) in water

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide

348270010 348270250 348271000
TS348270010EA 183 USD
TS348270010 TS348270250 TS348271000
N-Methylmorpholine-N-oxide 50% (w/w) in water
N-Methylmorpholine-N-oxide
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: –20 °C
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005947
CAS Number: 7529-22-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Strength 49 to 51 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR