Ammonium trifluoroacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammoniumtrifluoroacetate, Trifluoroacetic acid ammonium salt

218860250 218861000
TS218860250EA 71 USD
TS218860250 TS218861000
Ammonium trifluoroacetate 98%
Ammonium trifluoroacetate
Formula: C₂H₄F₃NO₂
MW: 131.05 g/mol
Melting Pt: 122…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00012615
CAS Number: 3336-58-1
EINECS: 222-072-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 122 to 126 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR